Learn More
RUBIS High Precision Tweezers
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
Specifications
Specifications
| Tip Style | Pointed, Ultra Fine |
| Design | Straight |
| Length (English) | 4.72 in. |
| Material | Anti-acid, Anti-magnetic Stainless Steel |
| Length (Metric) | 120 mm |
| Product Type | High Precision Tweezers |
| Quantity (No. per Case) | Each |
| Precision | Yes |
| Type | Tweezers, Precision |