missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548291
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
STARD6 Polyclonal antibody specifically detects STARD6 in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
STARD6 | |
Polyclonal | |
Unconjugated | |
STARD6 | |
StAR related lipid transfer domain containing 6; STARD6; StAR-related lipid transfer (START) domain containing 6; StAR-related lipid transfer domain containing 6; stAR-related lipid transfer protein 6; START domain containing 6; START domain-containing protein 6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
147323 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
P59095 | |
STARD6 | |
KLH conjugated synthetic peptide between 187-215 amino acids from the C-terminal region of human STARD6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |