missing translation for 'onlineSavingsMsg'
Learn More

STAT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 51070-2-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886355

  • 329.85 € / 150 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The STAT 4 protein is a transcription factor activated by hormone and cytokines. STAT 4 is most similar to STAT 1 (52%) and STAT 3 (47%). Functionally, STAT 4 is similar to other STAT family members in that it can be tyrosine phosphorylated by JAK 1 or JAK 2. STAT 4 forms homodimers and heterodimers with related STAT family members. Tyrosine phosphorylated STAT 4 can bind the interferon g activated site (GAS), and serine phosphorylation is also required for maximal transcriptional activity. STAT 4 expression is restricted to the thymus, spleen, and testis.
Spezifikation

Spezifikation

STAT4
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P42228, Q14765
SLEB11, STAT4
Rabbit
IgG
150 μL
-20°C
Primary
20849, 367264, 6775
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
STAT4
Liquid
Stat4
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation