missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAT5 alpha Monoclonal Antibody (ST5a-2H2)
GREENER_CHOICE

Artikelnummer. 10352843
Change view
Click to view available options
Menge:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
10352843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 10352843 Lieferant Invitrogen™ Lieferanten-Nr. 133600

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Mouse Monoclonal Antibody

This antibody recognizes the STAT5a protein from a variety of species including human, mouse, and rat and it reacts specifically with the STAT5a protein and does not exhibit any cross-reactivity with the highly related STAT5b protein. Cross-reactivity with other endogenous STAT isoforms has not been observed. Reactivity has been confirmed by western blotting using cell lysates derived from human A431 cells (+/- EGF treatment), NIH 3T3 cells, MCF-7 cells, and HeLa cells.

STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen STAT5 alpha
Anwendungen ELISA, Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon ST5a-2H2
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.1% sodium azide; pH 7.4
Gen STAT5A
Gen-Zugriffsnummer P42229, P42230, Q62771
Gen-Alias AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5
Gensymbole STAT5A
Wirtsspezies Mouse
Immunogen A peptide corresponding to the C-terminus of murine Stat5a coupled to a protein carrier.
Reinigungsverfahren Affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 20850, 24918, 6776
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG1 κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.