missing translation for 'onlineSavingsMsg'
Learn More

Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate, Mikromol™

Product Code. 17987124
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987124 25 mg 25mg
17939224 100 mg 100mg
30458727 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17987124 Supplier Mikromol Supplier No. MM1290.030025
 Restricted Item
Verfügbar ab: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate
CAS 32643-00-8
Analyte or Component Names Butyl 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoate
Shipping Condition Room Temperature
Molecular Formula C21 H28 N2 O5 S
Certifications/Compliance ISO 17025
Synonym Butyl 3-(Butylamino)-4-phenoxy-5-sulphamoylbenzoate, Bumetanide Imp. C (EP), Benzoic acid, 3-(aminosulfonyl)-5-(butylamino)-4-phenoxy-, butyl ester, Benzoic acid, 3-(butylamino)-4-phenoxy-5-sulfamoyl-, butyl ester (8CI), PF 1760, Bumetanide Impurity C
InChI Formula InChI=1S/C21H28N2O5S/c1-3-5-12-23-18-14-16(21(24)27-13-6-4-2)15-19(29(22,25)26)20(18)28-17-10-8-7-9-11-17/h7-11,14-15,23H,3-6,12-13H2,1-2H3,(H2,22,25,26)
SMILES CCCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)OCCCC
IUPAC Name butyl 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoate
Molecular Weight (g/mol) 420.52
Quantity 25 mg
Formula Weight 420.1719 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.