Learn More
Sartorius™ X100 NPS WORT 50MM ,45μ STERILE
Artikelnummer. 11700686 Alle ansehen Sartorius Produkte
Change view
Click to view available options
:
Durchmesser 50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
11700686 Durchmesser 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11700686 Lieferant Sartorius™ Lieferanten-Nr. 1409250RDN