Learn More
Autoklavier-Indikatorband
For use in high vacuum, high temperature steam autoclaves and other sterilisation systems.
Marke: Synergy Healthcare 31002550VE
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
- Manufactured from the same materials as standard pouches but with a self sealing adhesive strip
- Simple removal of the backing strip, folding over and firm pressure will ensure a correct seal