Learn More
ibidi sticky-Slide 8 Well High
Product Code. 31940947
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
90/Cs.
Unit Size:
Pack of 120
Pack of 90
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case Unit Size
31940947 90/Cs. Pack of 90
31940982 120/Cs. Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31940947 Supplier ibidi Supplier No. 8080890
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
An 8 well bottomless μ-Slide with high walls and a self-adhesive underside.
  • Sterilized
Specifications
Type Chamber Slide
Wells 8
No. per Case 90/Cs.
No. per Pack 1 Ea.
Sterility Sterile