Learn More
Ibidi Chemotaxis Sticky-Slide
Chemotaxis Sticky-Slide is a ready-to-use bottomless slide for chemotaxis assays with fast or slow migrating cells with a self-adhesive underside to which own substrates can be mounted.
Marke:  Ibidi 80328
Weitere Details : Netto-Gewicht : 0.15100kg
Artikelnummer. 17216173
  • 507.60 € / 10 Stück
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • A ready-to-use microscopy slide for chemotaxis assays with fast or slow migrating cells
  • suitable for 2D or 3D cell culture
  • stable chemotactic gradients for long-term experiments
  • bottomless with self-adhesive underside
  • sterilized
Spezifikation
Spezifikation
Microscope Slide
Sterile
10
1
Chemotaxis Sticky Slide