Learn More
Ibidi Sticky-Slide I Luer
Sticky-Slide I Luer is a bottomless channel slide used for perfusion applications with a self-adhesive underside.
Marke: Ibidi 80168
Weitere Details : Netto-Gewicht : 0.15800kg
Beschreibung
- A slide with one channel for cell cultivation under flow and for applying defined shear stress
- easy to connect with tubing and pump systems via female Luers
- bottomless with self-adhesive underside
- sterilized
Spezifikation
Microscope Slide | |
Sterile | |
200 μm | |
15 |