missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16223-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
STK32A (serine/threonine kinase 32A), also known as YANK1, is a 396 amino acid protein that belongs to the superfamily of serine/threonine protein kinases and exists as three isoforms. The gene encoding STK32A maps to human chromosome 5, which is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene.Spezifikation
STK32A | |
Polyclonal | |
Unconjugated | |
STK32A | |
serine/threonine kinase 32A, STK32A, YANK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
202374, 269019, 364858 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGW6, Q8WU08 | |
Stk32a | |
STK32A Fusion Protein Ag9276 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |