missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK33 Recombinant Human Protein
STK33 is a serine/threonine kinase.
Marke: Invitrogen™ PR7986A
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
STK33 is involved in signal transduction and cellular communication.
Spezifikation
NP_112168 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
STK33 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
61.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMK Unique Kinase Family | |
Recombinant | |
Baculovirus |