Learn More
Invitrogen™ STMN2 Polyclonal Antibody
Beschreibung
STMN2 Polyclonal Antibody for Western Blot, IHC (F), ICC/IF
STMN2, also known as SCG-10, is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. STMN2 may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. Reductions in the expression of this gene have been associated with Down's syndrome and Alzheimer's disease.
Spezifikation
Spezifikation
| Antigen | STMN2 |
| Anwendungen | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.02% sodium azide |
| Gen | Stmn2 |
| Gen-Zugriffsnummer | P21818, P55821, Q90987, Q93045 |
| Gen-Alias | AI159727; hypothetical protein LOC534991; neuronal growth-associated protein (silencer element); neuron-specific growth-associated protein; Protein SCG10; SCG10; Scgn10; SGC10; stathmin 2; stathmin-2; stathmin-like 2; Stmb2; Stmn2; superior cervical ganglia, neural specific 10; superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10 |
| Gensymbole | Stmn2 |
| Mehr anzeigen |