Learn More
STMND1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA566968
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: AEVAFAKGLQR VRSAGFEPSD LQGGKPLKRK KSKCDATLID RNESDESFGV VESDMSYNQA Highest antigen sequence identity to the following orthologs - mouse 51%, rat 49%.
STMND1 gene ontology annotations related to this gene include microtubule depolymerization; neuron projection development; regulation of cytoskeleton organization; regulation of microtubule polymerization or depolymerization.Spezifikation
STMND1 | |
Polyclonal | |
Unconjugated | |
STMND1 | |
stathmin domain containing 1; stathmin domain-containing protein 1; STMND1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
401236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
H3BQB6 | |
STMND1 | |
Recombinant Human STMND1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only