missing translation for 'onlineSavingsMsg'
Learn More

Strontium, Plasma-Standardlösung, Sr 1000 μg/ml, Thermo Scientific Chemicals, Specpure™

Catalog Number p-4824966
Click to view available options
Menge:
50 mL
100 mL
500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Sr(NO3)2 in 5% HNO3
MFCD00134060
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Sr(NO3)2 in 5% HNO3
MFCD00134060

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Strontium, plasma standard solution
Namenshinweis Sr 1000μg/mL
Menge 50 mL
Concentration or Composition (by Analyte or Components) Salpetersäure:5 %; Strontiumnitrat:0,24%; Wasser:94,76%
Dampfdruck 23 hPa (17mm Hg) at 20°C
Physikalische Form Flüssig

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'