Learn More
R&D Systems™ Substance P Parameter Assay Kit, R&D Systems™
a 3.5 hour competitive enzyme immunoassay designed to measure Substance P in cell culture supernates, serum, plasma, saliva, and urine.
Marke:  R&D Systems™ KGE007
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418055
  • 555.12 € / 1 Set
Voraussichtlicher Liefertermin 24-01-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
It contains a synthetically derived human Substance P peptide and has been shown to accurately quantitate this peptide. Results obtained using samples containing natural Substance P showed linear curves that were parallel to the standard curves obtained using the Parameter kit standards. These results indicate that this kit can be used to determine relative mass values for natural Substance P
Spezifikation
Spezifikation
39.00 - 2500pg/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
ELISA kit
43.8pg/mL
Multi-species
Biotin
ELISA
1 kit
Cell Culture Supernates (50uL), Serum (50uL), Heparin Plasma (50uL), Saliva (50uL), Urine (50uL)
1 x 96-well plate
Videos