missing translation for 'onlineSavingsMsg'
Learn More

Succinic anhydride, MilliporeSigma™

Product Code. 10220231
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10220231 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10220231 Supplier Sigma Aldrich Supplier No. 8006830100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

Specifications

pH 2.7 (20g/L, aq.)
Melting Point 116°C to 120°C
CAS 108-30-5
Color Colorless
Density 1.48g/cm3
Quantity 100 g
Odor Stinging
Packaging Plastic Bottle
Physical Form Crystals
Molecular Formula C4H4O3
MDL Number MFCD00005525
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
Boiling Point 261°C
Solubility Information Water: 67g/L at 20°C Hydrolysis
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
SMILES O=C1CCC(=O)O1
IUPAC Name oxolane-2,5-dione
Molecular Weight (g/mol) 100.07
PubChem CID 7922
ChEBI CHEBI:36595
Formula Weight 100.07
Chemical Name or Material Succinic anhydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.