missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
SUCLG1 Polyclonal specifically detects SUCLG1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
Spezifikation
| Antigen | SUCLG1 |
| Anwendungen | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Alexa Fluor 594 |
| Zusammensetzung | 50 mM Sodium Borate |
| Gen-Alias | EC 6.2.1, EC 6.2.1.4, FLJ21114, FLJ43513, GALPHA, MTDPS9, SCS-alpha, succinate-CoA ligase, alpha subunit, succinate-CoA ligase, GDP-forming, alpha subunit, succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial, Succinyl-CoA synthetase subunit alpha, SUCLA1 |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human SUCLG1 fragment. |
| Reinigungsverfahren | Antigen Affinity-purified |
| Menge | 0.1 mL |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?