missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilsäure-d4, TRC

Artikelnummer. 30426160
Change view
Click to view available options
Menge:
10 mg
Packungsgröße:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30426160 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426160 Supplier Toronto Research Chemicals Supplier No. TRCS68909710MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Sulfanilic Acid-d4
CAS 1235219-21-2
Summenformel C6H3D4NO3S
Menge 10 mg
Synonym 4-Amino(benzene-d4)sulfonic Acid, 1-Amino-(benzene-d4)-4-sulfonic Acid, 4-Aminophenylsulfonic Acid-d4, 4-Sulfoaniline-d4, (Aniline-d4)-4-sulfonic Acid, (Aniline-d4)-p-sulfonic Acid, NSC 7170-d4, Sulphanilic Acid-d4, p-Aminobenzenesulfonic Acid-d4, p-Aminophenylsulfonic Acid-d4, p-Sulfoaniline-d4
InChI-Formel InChI=1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)/i1D,2D,3D,4D
SMILES NC1=C([2H])C([2H])=C(S(=O)(O)=O)C([2H])=C1[2H]
IUPAC-Name 4-Aminobenzol-2,3,5,6-d4-sulfonsäure
Molekulargewicht (g/mol) 177.21
Formelmasse 177.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.