Learn More
SUPT7L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558841
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HEYCLKFTKL LRFAVDREAR LGQTPFPDVM EQVFHEVGIG SVLSLQKFWQ HRIKDYHSYM LQISKQLSEE YERIVNPEKA TEDAKPVKIK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
SUPT7L Polyclonal antibody specifically detects SUPT7L in Human samples. It is validated for Immunohistochemistry.Spezifikation
SUPT7L | |
Polyclonal | |
Unconjugated | |
SUPT7L | |
2610524B01Rik; 6030455L14Rik; Adenocarcinoma antigen ART1; KIAA0764; RGD1562206; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPT7L; SPT7-like STAGA complex gamma subunit; SPTF-associated factor 65 gamma; STAF65; STAF65(gamma); STAF65G; STAF65gamma; STAGA complex 65 gamma subunit; STAGA complex 65 subunit gamma; STAGA complex 65 subunit gamma-like; suppressor of Ty 7 (S. cerevisiae)-like; suppressor of Ty 7-like; SUPT7H; SUPT7L; Unknown (protein for MGC:137593) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9913 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94864 | |
SUPT7L | |
Recombinant protein corresponding to Human SUPT7L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only