Learn More
SUV39H2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563450
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NTVDLEGPPS DFYYINEYKP APGISLVNEA TFGCSCTDCF FQKCCPAEAG VLLAYNKNQQ IKIPPGTPI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
The murine gene Suv39h2 encodes an H3 histone methyltransferase (HMTase) 59% identical in sequence to mouse Suv39h1. During embryogenesis, both proteins overlap in tissue expression, yet Suv39h2 transcripts are restricted to the testes in adult animals. Immunolocalization of the Suv39h2 protein during spermatogenesis indicates enrichment at the heterochromatin from the leptotene to the round spermatid stage. Moreover, Suv39h2 specifically accumulates with chromatin of the sex chromosomes, which undergo transcriptional silencing during the first meiotic prophase. Suv39h2 HMTase may also organize meiotic heterochromatin with the potential for epigenetic imprint to the male germline.Spezifikation
SUV39H2 | |
Polyclonal | |
Unconjugated | |
SUV39H2 | |
4930507K23Rik; AA536750; D030054H19Rik; D2Ertd544e; FLJ23414; H3 K9 HMTase 2; H3-K9-HMTase 2; Histone H3-K9 methyltransferase 2; histone-lysine N-methyltransferase SUV39H2; KMT1; KMT1B; lysine N-methyltransferase 1B; RP11-2K17.2; Su; Su(var)3-9 homolog 2; suppressor of variegation 3-9 homolog 2; suppressor of variegation 3-9 homolog 2 (Drosophila); Suv39h histone methyltransferase; Suv39h2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79723 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H5I1 | |
SUV39H2 | |
Recombinant protein corresponding to Human SUV39H2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only