missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550578
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total SV2A protein.
SV2A Polyclonal antibody specifically detects SV2A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
SV2A | |
Polyclonal | |
Unconjugated | |
SV2A | |
AI746429; calcium regulator SV2A; Kiaa0736; mKIAA0736; PSEC0174; SV2; SV2A; synaptic vesicle glycoprotein 2 a; synaptic vesicle glycoprotein 2a; Synaptic vesicle protein 2; synaptic vesicle protein 2A; synaptic vesicle transmembrane transporter (SV2) RNA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117559, 64051, 9900 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q02563, Q7L0J3, Q9JIS5 | |
SV2A | |
Synthetic peptide corresponding to a region derived from internal residues of human synaptic vesicle glycoprotein 2A | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |