Learn More
SVEP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554245
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NRLDYSYDDF LDTVQETATS IGNAKSSRIK RSAPLSDYKI KLIFNITASV PLPDERNDTL EWENQQRLLQ TLETITNKLK RTLNKDPMYS FQLASEIL Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.
SVEP1 Polyclonal antibody specifically detects SVEP1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
SVEP1 | |
Polyclonal | |
Unconjugated | |
SVEP1 | |
C9orf13; CCP module-containing protein 22; CCP22; Polydom; selectin-like osteoblast-derived protein; selectin-like protein; SELOB; SEL-OB; Serologically defined breast cancer antigen NY-BR-38; sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1; sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1; SVEP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4LDE5 | |
SVEP1 | |
Recombinant protein corresponding to Human SVEP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only