Learn More
SVIL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554145
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LEGIENDTQP ILLQSCTGLV THRLLEEDTP RYMRASDPAS PHIGRSNEEE ETSDSSLEKQ TRSKYCTETS GVHGDSPYGS GTMDTHSLES KAERIARYKA ERRRQLAEKY GLTLDPEADS EYLSRYTKSR KEPDAVEKR Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
SVIL Polyclonal antibody specifically detects SVIL in Human samples. It is validated for Immunohistochemistry.Spezifikation
SVIL | |
Polyclonal | |
Unconjugated | |
SVIL | |
Archvillin; AU024053; B430302E16Rik; membrane-associated F-actin binding protein p205; p205/p250; Supervillin; supervillin muscle-specific isoform; Svil | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6840 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95425 | |
SVIL | |
Recombinant protein corresponding to Human SVIL | |
100 μL | |
Primary | |
Human | |
Antibody | |