Learn More
SYCE1 Monoclonal Antibody (6G7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524504
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Peptide Sequence: MLQECKERIS ALNLQIEEEK NKQRQLRLAF EEQLEDLMGQ HKDLWDFHMP ERLAKEICAL DSSKEQLLKE EKLVKATLED VKHQLCSLCG AEGPSTLDEG LFLRSQEAAA TVQLFQEEHR KAEELLAAAA QRHQQLQQKC QQQQQKRQRL KEELEKHGMQ VPAQAQSTQE EEAGPGDVAP RPGRPVTWWS
SYCE1 Monoclonal antibody specifically detects SYCE1 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
SYCE1 | |
Monoclonal | |
Unconjugated | |
SYCE1 | |
4933406J07Rik; C10orf94; Cancer/testis antigen 76; CT76; POF12; SPGF15; SYCE1; Synaptonemal complex central element protein 1; telomeric protein | |
Mouse | |
Protein A | |
RUO | |
93426 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
6G7 | |
PBS with no preservative; pH 7.4 | |
Q8N0S2 | |
SYCE1 | |
SYCE1 (NP_963858.1, 1 a.a. ∽ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |