Learn More
Synaptophysin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. 16857764 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16857764 20 μL 20 Mikroliter
16847764 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16857764 Lieferant Proteintech Lieferanten-Nr. 177851AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
Spezifikation
Antigen Synaptophysin
Anwendungen Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 0.25 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen SYP
Gen-Zugriffsnummer P07825, P08247, Q62277
Gen-Alias SYN, synaptophysin, SYP
Gensymbole SYP
Wirtsspezies Rabbit
Immunogen Synaptophysin Fusion Protein Ag11781
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 20977, 24804, 6855
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG