Learn More
SynGAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558362
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LHLYRDSDKK RKKDKAGYVG LVTVPVATLA GRHFTEQWYP VTLPTGSGGS GGMGSGGGGG SGGGSGGKGK GGCPAVRLKA RYQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
SynGAP Polyclonal antibody specifically detects SynGAP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
SynGAP | |
Polyclonal | |
Unconjugated | |
SYNGAP1 | |
DKFZp761G1421; Gm1963; KIAA1938; MRD5; Neuronal R; Neuronal RasGAP; OTTHUMP00000209046; p135 SynGAP; Ras GTPase-activating protein SynGAP; ras/Rap GTPase-activating protein SynGAP; RASA1; RASA5; synaptic Ras GTPase activating protein 1; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein 1 homolog (rat); synaptic Ras GTPase activating protein, 135kDa; synaptic Ras GTPase-activating protein 1; Synaptic Ras-GAP 1; SYNGAP; Syngap1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8831 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PV0 | |
SYNGAP1 | |
Recombinant protein corresponding to Human SynGAP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only