missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNGR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563698
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FQDLRNDAPV PYKRFLDEGG MVLTTL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. SYNGR3 is predominantly expressed in brain and placenta. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear.Spezifikation
SYNGR4 | |
Polyclonal | |
Unconjugated | |
Syngr4 | |
1700016O14Rik; MGC125805; synaptogyrin 4; synaptogyrin 4 protein; synaptogyrin-4; SYNGR4; testis tissue sperm-binding protein Li 72n | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23546 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95473 | |
Syngr4 | |
Recombinant protein corresponding to Human SYNGR4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only