Learn More
SYNJ2BP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551471
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NGRVDYLVTE EEINLTRGPS GLGFNIVGGT DQQYVSNDSG IYVSRIKENG AAALDGRLQE GDKILSVNGQ DLKNLLHQDA VDLFRNAGYA VSLRVQHRLQ VQNGPIGHRG EG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
SYNJ2BP Polyclonal antibody specifically detects SYNJ2BP in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SYNJ2BP | |
Polyclonal | |
Unconjugated | |
SYNJ2BP | |
AA409442; activin receptor interacting protein 2; activin receptor interacting protein 5; activin receptor-interacting protein 2; activin receptor-interacting protein 2a; activin receptor-interacting protein 4; ActRIP4; Arip2; ARIP2a; D12Wsu118e; Mitochondrial outer membrane protein 25; NPW16; Omp25; outer membrane protein; outer membrane protein 25; synaptojanin 2 binding protein; synaptojanin-2-binding protein; SYNJ2BP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24071, 55333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57105, Q9D6K5 | |
SYNJ2BP | |
Recombinant protein corresponding to Human SYNJ2BP | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only