Learn More
SYNPO2L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563082
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PAEALLLPHG PLRPGPHLIP MVGPVPHPVA EDLTTTYTQK AKQAKLQRAE SLQEKSIKEA KTKCRTIASL LTA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
SYNPO2L was initially identified as a novel heart-enriched gene that encodes a cytoskeletal protein highly expressed in the Z-disc of heart and skeletal muscle, associates with actin and interacts with a-actinin. It is a member of the synaptopodin family, sharing greatest homology with Synaptopodin 2. Recent studies have shown that SYNPO2L, while primarily localized to the sarcomere, can also translocate to the nucleus. A knockdown of SYNPO2L in zebrafish resulted in aberrant cardiac and skeletal muscle development and function, suggesting that it is a critical component of the sarcomere and plays an important role in muscle development.Spezifikation
SYNPO2L | |
Polyclonal | |
Unconjugated | |
SYNPO2L | |
1110054M18Rik; Chap; RGD1565434; synaptopodin 2 like; synaptopodin 2-like; synaptopodin 2-like protein; SYNPO2L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79933 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H987 | |
SYNPO2L | |
Recombinant protein corresponding to Human SYNPO2L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only