missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYPL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553126
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ATGHNIIDEL PPCKKKAVLC YFGSVTSMGS Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 70%.
SYPL1 Polyclonal antibody specifically detects SYPL1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
SYPL1 | |
Polyclonal | |
Unconjugated | |
SYPL1 | |
AI314763; AI604763; D12Ertd446e; H-SP1; Pan I; PanI; Pantophysin; Pphn; synaptophysin like 1; synaptophysin-like 1; synaptophysin-like protein; synaptophysin-like protein 1; SYPL; Sypl1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6856 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16563 | |
SYPL1 | |
Recombinant protein corresponding to Human SYPL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only