missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550271
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total SYT17 protein.
SYT17 Polyclonal antibody specifically detects SYT17 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SYT17 | |
Polyclonal | |
Unconjugated | |
SYT17 | |
B/K protein; Bk; brain and kidney protein; protein B/K; synaptotagmin 17; synaptotagmin XVII; Synaptotagmin-17; Syt17; SytXVII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110058, 51760 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q920M7, Q9BSW7 | |
SYT17 | |
Fusion protein corresponding to residues near the N terminal of human synaptotagmin XVII | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |