missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550268
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total SYT3 protein.
Synaptotagmin 3 Polyclonal antibody specifically detects Synaptotagmin 3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Synaptotagmin 3 | |
Polyclonal | |
Unconjugated | |
SYT3 | |
AI385753; SIII; synaptotagmin 3; synaptotagmin III; Synaptotagmin3; Synaptotagmin-3; SynaptotagminIII; Syt III; Syt3; SYT-3; SytIII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84258 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BQG1 | |
SYT3 | |
Fusion protein corresponding to a region derived from internal residues of human synaptotagmin III | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |