missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin 7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550270
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total SYT7 protein.
Synaptotagmin 7 Polyclonal antibody specifically detects Synaptotagmin 7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Synaptotagmin 7 | |
Polyclonal | |
Unconjugated | |
SYT7 | |
AI851541; B230112P13Rik; IPCA7; IPCA-7; PCANAP7; Prostate cancer-associated protein 7; Protein Syt7; synaptotagmin 7; synaptotagmin VII; Synaptotagmin-7; SYT7; SYT-7; SytVII; SYT-VII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9066 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O43581 | |
SYT7 | |
Fusion protein corresponding to a region derived from internal residues of human synaptotagmin VII | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |