Learn More
SYTL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551605
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RLKNELLEIK RKGAKRGSQH YSDRTCARCQ ESLGRLSPKT NTCRGCNHLV CRDCRIQESN GTWRCKVCAK EIELKKATGD WFYDQKVNRF AYRTGSEIIR MSLRHKPAVS KRETVGQSLL HQTQMGDIWP GRKIIQERQK EPSVLFEVP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
SYTL4 Polyclonal antibody specifically detects SYTL4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SYTL4 | |
Polyclonal | |
Unconjugated | |
SYTL4 | |
Exophilin-2; Gph; Granuphilin; granuphilin b; granuphilin-a; granuphilin-b; Slp4; synaptotagmin like 4; synaptotagmin-like 4; synaptotagmin-like 4 (granuphilin-a); synaptotagmin-like protein 4; Sytl4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94121 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96C24 | |
SYTL4 | |
Recombinant protein corresponding to Human SYTL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only