Learn More
Descrizione
T-Lab Eco:
Affordable Performance and Trusted Reliability for your laboratory sterilization needs. T-Lab Eco was developed especially for educational institutions and research facilities. Designed with simplicity and economy in mind. Featuring the only IoT software designed for the smart laboratory, connecting sterilization devices and communicating on-site or remotely. Cloud based and paperless. This vertical floor-standing autoclave covers the fundamental needs for general lab sterilization with the aim of increasing the productivity of your laboratory. Offering universal capacity, together with the optimization of resources such as water, power and operating time results. Intended for the sterilization of a wide range of liquids and solids such as glassware, metal utensils and other laboratory items.
- Reliable Performance - Solid, glassware and liquid loads. Efficient sterilization time including cooling.
- Affordable - Built with just the essential features. Less water wasted, less electricity consumed, less time spent on maintenance. Cylindrical chamber is less expensive and more reliable over a long working life.
- Plug and Play Installation - This autoclave does not require dedicated installation connections, simply a power source.
- Versatile for all your basic needs - Sterilizes liquids, solids, sealed and unsealed bottles. Performance and quality within a small space.
- Unparalleled User Experience - High resolution multicolour touchscreen which adjustable screen colours. Monitor your autoclave connected to a network with R.PC.R. software.
- Standard Features - Two PT100 sensors. Built-in cooling fan. Built-in memory: 1000 cycles.
Specifica
Specifica
| Capacity (English) | 13.2 gal. |
| Capacity (Metric) | 60 L |
| Depth (Metric) Chamber | 50.5 cm |
| Diameter (English) Chamber | 15.6 in. |
| Depth (English) Chamber | 20.2 in. |
| Diameter (Metric) Chamber | 39 cm |
| Length (English) | 32.4 in. |
| Length (Metric) | 81 cm |
| Product Type | Autoclave, Top-loading |
| Temperature (English) | 221°F to 278°F |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.