Learn More
TACC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556617
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NLPTHGGQEQ ALGSELQSQL PKGTLSDTPT SSPTDMVWES SLTEESELSA PTRQKLPALG EKRPEGACGD GQSSRVSPPA ADVLKDFSLA GNFS Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 41%.
TACC2 Polyclonal antibody specifically detects TACC2 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TACC2 | |
Polyclonal | |
Unconjugated | |
TACC2 | |
anti zuai-1; Anti-Zuai-1; AZU-1; ECTACC; mKIAA4180; MNCb-3527; RP11-296H2.1; TACC2; transforming acidic coiled coil 2; transforming acidic coiled-coil containing protein 2; transforming acidic coiled-coil containing protein 2 variant 3; transforming acidic coiled-coil-containing protein 2; transforming, acidic coiled-coil containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10579 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95359 | |
TACC2 | |
Recombinant protein corresponding to Human TACC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only