missing translation for 'onlineSavingsMsg'
Learn More

TAF12 Monoclonal Antibody (22TA-2A1), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA3072

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15749230

  • 452.00 € / 50 Mikroliter
Verfügbar ab: 24-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

MA3-072 detects TATA-Binding Protein (TBP) Associated factor 12 in the human samples. MA3-072 has been successfully used in Western blot, immunofluorescence, and flow cytometry applications. The MA3-072 immunogen is a recombinant human TAF12 protein. Epitope is localized at the C-terminal part of TAF12.

TAF12 Monoclonal antibody specifically detects TAF12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

TAF12
Monoclonal
Conc. Not Determined
ascites with 0.05% sodium azide
Q16514
TAF12
Recombinant human TAF12 protein. Epitope localized at the C-terminal part of TAF12.
RUO
6883
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Western Blot
22TA-2A1
Unconjugated
TAF12
20kDa; 2810422D08Rik; AW557038; hypothetical protein LOC614227; TAF12; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kD; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kDa; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa; TAF15; TAF2J; TAFII20; TAFII-20; TAFII20/TAFII15; TAFII-20/TAFII-15; TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD; TATA box binding protein associated factor 12; TATA-box binding protein associated factor 12; TATA-box binding protein associated factor 12; transcription initiation factor TFIID subunit 12; transcription initiation factor TFIID 20 kDa subunits; Transcription initiation factor TFIID 20/15 kDa subunits; transcription initiation factor TFIID subunit 12
Mouse
50 μL
Primary
Human
Antibody
IgG1 κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only