Learn More
TAF13 Monoclonal Antibody (16TA-3C12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA3073
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA3-073 detects TATA Binding protein (TBP) Associated factor at 14kDa in the human and mouse samples. MA3-073 has been succesfully used in Western blot application. The MA3-073 immunogen is a human recombinant human TAF13 protein.
TAF13 Monoclonal antibody specifically detects TAF13 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
TAF13 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.05% sodium azide | |
P61216, Q15543 | |
TAF13 | |
Recombinant human TAF13 protein | |
RUO | |
6884, 99730 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
16TA-3C12 | |
Unconjugated | |
TAF13 | |
1810004N01Rik; 2010309N11Rik; AI847295; MGC22425; TAF(II)18; TAF13; TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 18kDa; TAF13 RNA polymerase II, TATA box-binding protein-associated factor; Taf2k; TAFII18; TAFII-18; TATA box binding protein (TBP)-associated factor, RNA polymerase II, K, 18kD; TATA box binding protein associated factor 13; TATA-box binding protein associated factor 13; TBP-associated factor, RNA polymerase II, K, 18kD; transcription initiation factor TFIID 18 kD subunit; transcription initiation factor TFIID 18 kDa subunit; Transcription initiation factor TFIID subunit 13 | |
Mouse | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |