Learn More
TAF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552310
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LSDIINTPLP RVPLDVCLKA HWLSIEGCQP AIPENPPPAP KEQQKAEATE PLKSAKPGQE EDGPLKGKGQ GATTADGKGK EKKAPPLLEG APLRLKPRSI HELSVEQQLY YKEITEACVG SCEAKRAEAL QSIATDPG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
TAF6 Polyclonal antibody specifically detects TAF6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TAF6 | |
Polyclonal | |
Unconjugated | |
TAF6 | |
80kDa; AW549759; DKFZp781E21155; MGC:8964; p80; RNA polymerase II TBP-associated factor subunit E; RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAF(II)70; TAF(II)80; TAF2E; Taf6; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAFII70; TAFII-70; TAFII80; TAFII-80; TAFII85; TATA box binding protein (Tbp)-associated factor, RNA polymerase II, E; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85kD; TATA box binding protein associated factor 6; TATA-box binding protein associated factor 6; TBP-associated factor 6; TBP-associated factor 6 isoform alpha; TFIID subunit p80; transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; transcription initiation factor TFIID subunit 6; wu:fb83e08; wu:fj30b0 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6878 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49848 | |
TAF6 | |
Recombinant protein corresponding to Human TAF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only