Learn More
Tocris Bioscience™ TAK 960 hydrochloride
Potent and selective PLK1 inhibitor
N.V.
Spezifikation
Produkttyp TAK 960 hydrochloride
Beschreibung TAK 960 hydrochloride
Molekulargewicht 598.06
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
Potent PLK1 inhibitor (IC50 values are 0.8, 16.9 and 50.2 nM for PLK1, PLK2 and PLK3 respectively). Exhibits >20-fold selectivity for PLK1 over FAK, MLCK and FES, and has minimal activity against a panel of 282 other kinases. Inhibits proliferation of a range of cancer cell lines in vitro and inhibits tumor growth of multiple human cancer cell xenografts in vivo. Orally bioavailable.
Spezifikation
Spezifikation
TAK 960 hydrochloride
TAK 960 hydrochloride
598.06