missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talkum, Pulver, Multikompendial, J.T.Baker™
Click to view available options
Menge:
500 g
2 kg
Verpackung:
Poly-Flasche
Spezifikation
Chemischer Name oder Material | Talkum |
Verpackung | Poly-Flasche |
Menge | 500 g |
Güte | Multi-Kompendium |
Physikalische Form | Pulver |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'