missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562509
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RHTRQMRNTV AGSRVPGRGA PISA Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 50%.
TAS2R1 Polyclonal antibody specifically detects TAS2R1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TAS2R1 | |
Polyclonal | |
Unconjugated | |
TAS2R1 | |
MGC126778; MGC126780; T2R1; TAS2R1; taste 2 receptor member 1; taste receptor family B member 7; Taste receptor type 2 member 1; taste receptor, family B, member 7; taste receptor, type 2, member 1; TRB7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYW7 | |
TAS2R1 | |
Recombinant protein corresponding to Human TAS2R1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only