missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R42 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557544
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SLVRHTRNLK LSSLGSRDSS TEAHRRAMK Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.
TAS2R42 Polyclonal antibody specifically detects TAS2R42 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TAS2R42 | |
Polyclonal | |
Unconjugated | |
TAS2R42 | |
candidate taste receptor hT2R55; hT2R55; T2R24; T2R42; T2R55; TAS2R42; TAS2R55; taste 2 receptor member 42; taste receptor type 2 member 42; taste receptor type 2 member 55; taste receptor, type 2, member 42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
353164 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTR8 | |
TAS2R42 | |
Recombinant protein corresponding to Human TAS2R42 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only