missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R42 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567770
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Mouse and Rat samples.
TAS2R42 Polyclonal antibody specifically detects TAS2R42 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
TAS2R42 | |
Polyclonal | |
Unconjugated | |
TAS2R42 | |
candidate taste receptor hT2R55; hT2R55; T2R24; T2R42; T2R55; TAS2R42; TAS2R55; taste 2 receptor member 42; taste receptor type 2 member 42; taste receptor type 2 member 55; taste receptor, type 2, member 42 | |
Rabbit | |
affinity chromatography | |
RUO | |
353164 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7RTR8 | |
TAS2R42 | |
Synthesized peptide derived from human TAS2R42. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |