missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Oligomeric Complex Antibody (TOC1), Alexa Fluor™ 350 - Chimeric, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Tau Oligomeric Complex Monoclonal antibody specifically detects Tau Oligomeric Complex in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Spezifikation
Spezifikation
| Antigen | Tau Oligomeric Complex |
| Anwendungen | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | TOC1 |
| Konjugat | Alexa Fluor 350 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | Tau Oligomeric Complex 1, TOC1, TOC-1 |
| Wirtsspezies | Rabbit |
| Immunogen | The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers. |
| Reinigungsverfahren | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?