missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAX1BP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14424-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a HTLV-1 tax1 binding protein. The encoded protein interacts with TNFAIP3, and inhibits TNF-induced apoptosis by mediating the TNFAIP3 anti-apoptotic activity. Degradation of this protein by caspase-3-like family proteins is associated with apoptosis induced by TNF. This protein may also have a role in the inhibition of inflammatory signaling pathways. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
TAX1BP1 | |
Polyclonal | |
Unconjugated | |
TAX1BP1 | |
CALCOCO3, T6BP, Tax1 binding protein 1, TAX1BP1, TRAF6 binding protein, TXBP151 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52440, 8887 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UKC1, Q86VP1 | |
Tax1bp1 | |
TAX1BP1 Fusion Protein Ag5789 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |