Learn More
Tazarotene, TRC
Product Code. 17962005
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962005 50 mg 50mg
17961795 10 mg 10mg
17993565 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17962005 Supplier Toronto Research Chemicals Supplier No. TRCT01005050MG
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118292-40-3
Molecular Formula C21 H21 N O2 S
Molecular Weight (g/mol) 351.46
Synonym Tazarotene, 3-Pyridinecarboxylic acid, 6-[2-(3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-, ethyl ester, 3-Pyridinecarboxylic acid, 6-[(3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-, ethyl ester (9CI), 2H-1-Benzothiopyran, 3-pyridinecarboxylic acid deriv., AGN 190168, Acnitaz, Tazarotene, Tazorac, Zorac
IUPAC Name ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
SMILES CCOC(=O)c1ccc(nc1)C#Cc2ccc3SCCC(C)(C)c3c2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C21H21NO2S/c1-4-24-20(23)16-7-9-17(22-14-16)8-5-15-6-10-19-18(13-15)21(2,3)11-12-25-19/h6-7,9-10,13-14H,4,11-12H2,1-3H3
Formula Weight 351.1293
Chemical Name or Material Tazarotene