missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D22A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550801
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total TBC1D22A protein.
TBC1D22A Polyclonal antibody specifically detects TBC1D22A in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
TBC1D22A | |
Polyclonal | |
Unconjugated | |
TBC1D22A | |
BC023106; C22orf4; D15Ertd781e; HSC79E021; putative GTPase activator; RGD1306588; TBC1 domain family member 22A; TBC1 domain family, member 22a; TBC1D22A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25771 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8WUA7 | |
TBC1D22A | |
Fusion protein of human TBC1 domain family, member 22A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |