Learn More
TBC1D8B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554911
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LHVSKPANEK EAESAKHSPE KGKGKIDIQA YLSQWQDELF KKEENIKDLP RMNQSQFIQF SKTLYNLFHE DPEEESLYQA IAVVTSLLLR MEEVGRKLHS PTSSAKGFSG TVCGSGGPSE EKTGSHLEKD PCSFREEPQW SFAFEQILA Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.
TBC1D8B Polyclonal antibody specifically detects TBC1D8B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TBC1D8B | |
Polyclonal | |
Unconjugated | |
TBC1D8B | |
4921505D17Rik; 9030605E16Rik; GRAMD8B; RGD1563084; RP11-321G1.1; TBC1 domain family member 8B; TBC1 domain family, member 8B; TBC1 domain family, member 8B (with GRAM domain); TBC1D8B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54885 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0IIM8 | |
TBC1D8B | |
Recombinant protein corresponding to Human TBC1D8B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only